Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS571

Sigma-Aldrich

Anti-3-PGDH Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

D-3-phosphoglycerate dehydrogenase, 3-PGDH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PHGDH(26227)

General description

3-PGDH (PHGDH) protein in an important enzyme in L-serine biosynthesis. 3-PGDH performs the first rate limiting step in the phosphorylated pathway of serine biosynthesis. 3-PGDH is also a biomarker in certain cancers that over express 3-PGDH. Moreover, 3-PGDH is the essential enzyme needed for cells to traverse the S phase and divide, and it plays a key role in T cell activation and proliferation for instance. Interestingly, even though 3-PGDH knockdown and reduced serine synthesis slows growth and proliferation of many cancer cells in vitro 3-PGDH knockdown does not affect tumor maintenance and growth in established breast cancer xenograft models, suggesting that 3-PGDH-dependent cancer cell growth may be context-dependent.

Immunogen

Epitope: Near N-terminus
KLH-conjugated linear peptide corresponding to human 3-PGDH near the N-terminus.

Application

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected 3-PGDH in human cerebral cortex, human kidney, and human cerebellum tissue.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
This Anti-3-PGDH Antibody is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of 3-PGDH.

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected 3-PGDH in 10 µg of A431 cell lysate.

Target description

~54 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Zhang et al.
eLife, 8 (2019-05-01)
The human macula is more susceptible than the peripheral retina to developing blinding conditions such as age-related macular degeneration, diabetic retinopathy. A key difference between them may be the nature of their Müller cells. We found primary cultured Müller cells
Weiyong Shen et al.
Glia, 69(8), 1966-1986 (2021-04-10)
The importance of Müller glia for retinal homeostasis suggests that they may have vulnerabilities that lead to retinal disease. Here, we studied the effect of selectively knocking down key metabolic genes in Müller glia on photoreceptor health. Immunostaining indicated that
Yanling Xin et al.
eLife, 11 (2022-12-03)
N 6-methyladenosine (m6A) is the most prevalent mRNA internal modification and has been shown to regulate the development, physiology, and pathology of various tissues. However, the functions of the m6A epitranscriptome in the visual system remain unclear. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service