Skip to Content
MilliporeSigma

ABS312

Anti-Fatty-acid binding protein 5 Antibody

from rabbit

Synonym(s):

Fatty acid-binding protein, epidermal, Epidermal-type fatty acid-binding protein, E-FABP, Fatty acid-binding protein 5, Psoriasis-associated fatty acid-binding protein homolog, PA-FABP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FABP5(2171)

General description

Fatty acid-binding protein 5 (FABP5) is also called Fatty acid-binding protein, epidermal, Epidermal-type fatty acid-binding protein (E-FABP), and Psoriasis-associated fatty acid-binding protein homolog (PA-FABP). FABP5 has a high specificity for fatty acids, particularly for C18 chain lengths. The addition of double bonds or decreasing chain lengths reduces FABP5 affinity. FABP5 is likely involved in keratinocyte differentiation, and it is expressed in keratinocytes, especially in psoriatic skin.
~ 16 kDa observed

Immunogen

Recombinant protein corresponding to human Fatty-acid binding protein 5.

Application

Detect Fatty Acid Binding Protein using this rabbit polyclonal antibody, Anti-Fatty-acid binding protein 5 Antibody validated for use in western blotting.
Research Category
Signaling
Research Sub Category
Lipid Metabolism & Weight Regulation
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected Fatty-acid binding protein 5 in 10 µg of NIH/3T3 (mouse) and PC12 (rat) cell lysate.

Physical form

Format: Purified
Protein G Purified
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Fatty-acid binding protein 5 in 10 µg of A431 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library