Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN903

Sigma-Aldrich

Anti-Huntingtin

from rabbit, purified by affinity chromatography

Synonym(s):

Huntington disease protein, HD protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... HTT(3064)

Related Categories

General description

Huntingtin (UniProt: P42858; also known as Huntington disease protein, HD protein) is encoded by the HTT (also known as HD, IT15) gene (Gene ID: 3064) in human. The protein is found in the perinuclear region along with microtubules, and in the centrosomal region along with gamma-tubulin. It is expressed in the brain and is mainly found in the cerebellar cortex, the neocortex, the striatum, and the hippocampal formation. Huntingtin is necessary for neuronal survival and is involved in synaptic vesicle trafficking, microtubule binding and may also have a role in apoptosis. It plays a role in microtubule-mediated transport and vesicle function. Huntingtin protein contains a nuclear export signal (aa 2395-2404) and 5 HEAT (Huntington, Elongation Factor 3, PR65/A, TOR) domains. Huntingtin protein is cleaved by apopain downstream of the polyglutamine stretch and the resulting N-terminal fragment is cytotoxic and provokes apoptosis. Phosphorylation at Serine 1179 and 1199 by CDK5 in response to DNA damage in nuclei of neurons protects neurons against polyglutamine expansion as well as DNA damage mediated toxicity. Mutations in HTT gene cause Huntington disease that is characterized by involuntary movements (chorea), general motor impairment, psychiatric disorders and dementia. Wild-type huntingtin and anti-huntingtin antibody reduces aggregation and cellular toxicity of the mutant huntingtin form in mammalian cell models of Huntington disease.

Specificity

This rabbit polyclonal antibody detects Huntington disease protein in human and mouse brain. It targets an epitope within 180 amino acids from the N-terminal region.

Immunogen

Epitope: N-terminus
GST/His-tagged recombinant fragment corresponding to 180 amino acids from the N-terminal region of human Huntington disease protein .

Application

Anti-Huntingtin, Cat. No. ABN903, is a highly specific rabbit polyclonal antibody that targets Huntington Disease Protein and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: 2 µg/mL from a representative lot detected Huntingtin in mouse brain tissue lysate.

Quality

Evaluated by Immunohistochemistry in human cerebral cortex and human huntington′s diseased brain tissues.

Immunohistochemistry Analysis: A 1:50-250 dilution of this antibody detected Huntingtin in human cerebral cortex and human Huntington′s diseased brain tissues.

Target description

347.60 kDa calculated.

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tanpreet Kaur et al.
Frontiers in neuroscience, 15, 766176-766176 (2021-12-21)
Mutations in the huntingtin gene (HTT) triggers aggregation of huntingtin protein (mHTT), which is the hallmark pathology of neurodegenerative Huntington's disease (HD). Development of a high affinity 18F radiotracer would enable the study of Huntington's disease pathology using a non-invasive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service