Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN161-I

Sigma-Aldrich

Anti-pan Neurexin-1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Neurexin-1, Neurexin-1-alpha, Neurexin I-alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Nrxn1(18189)

General description

Neurexin-1 (UniProt Q9CS84; also known as Neurexin I-alpha, Neurexin-1-alpha) is encoded by the Nrxn1 (also known as Kiaa0578) gene (Gene ID 18189) in murine species. Neurexin (NRXN) is a presynaptic single pass transmembrane protein that assists in joining neurons together at the synapse. Neurexins mediate signaling across the synapse and neural networks by specifying synpatic function. Mutations in the genes encoding for neurexins are implicated in autism, schizphrenia, and nicotine dependence. There are two classifications of neurexins, α-NRXNs and β-NRXNs. The α-NRXNs are larger and have different amino-terminal extracellular sequences than β-NRXNs. β-NRXNs act as receptors for neuroligin and have been found to play a role in angiogenesis.

Specificity

Expected to react with all neurexin-1-alpha and neurexin-1-beta spliced isoforms of murine, human, and rat species, as this polyclonal antibody targets the C-terminal end cytoplasmic sequence that is not affected by alternative splicings (Missler, M., and Südhof, T.C. (1998). Trends Genet. 14(1):20-26). Cross-reactivities toward alpha and beta isoforms of neurexin-2 and neurexin-3 are also likely, although the C-termminal sequence is not 100% conserved among neurexin-1, neurexin-2 and neurexin-3.

Immunogen

Epitope: Near C-terminus.
KLH-conjugated linear peptide corresponding to a cytoplasmic domain sequence near the C-teminus of mouse neurexin-1.

Application

Anti-pan Neurexin-1 Antibody detects level of pan Neurexin-1 and has been published and validated for use in Western Blotting, Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Neurexin-1 in mouse, rat, and human cerebral cortex tissue sections.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected Neurexin-1 in 10 µg of mouse brain tissue lysate.

Target description

~170/55/40 kDa observed. Varible depending on the isoform(s) present in the test sample(s).

Linkage

Replaces: ABN161

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth Ann Roundhill et al.
Cellular oncology (Dordrecht), 44(5), 1065-1085 (2021-08-18)
The development of biomarkers and molecularly targeted therapies for patients with Ewing sarcoma (ES) in order to minimise morbidity and improve outcome is urgently needed. Here, we set out to isolate and characterise patient-derived ES primary cell cultures and daughter
Ralda Nehme et al.
Nature communications, 13(1), 3690-3690 (2022-06-28)
It is unclear how the 22q11.2 deletion predisposes to psychiatric disease. To study this, we generated induced pluripotent stem cells from deletion carriers and controls and utilized CRISPR/Cas9 to introduce the heterozygous deletion into a control cell line. Here, we
Sebnem Ece Eksi et al.
Nature communications, 12(1), 7292-7292 (2021-12-17)
Identifying precise molecular subtypes attributable to specific stages of localized prostate cancer has proven difficult due to high levels of heterogeneity. Bulk assays represent a population-average, which mask the heterogeneity that exists at the single-cell level. In this work, we
Alessandra Sclip et al.
Nature communications, 14(1), 4976-4976 (2023-08-18)
Synaptic adhesion molecules (SAMs) shape the structural and functional properties of synapses and thereby control the information processing power of neural circuits. SAMs are broadly expressed in the brain, suggesting that they may instruct synapse formation and specification via a
Nicolas Chofflet et al.
Frontiers in molecular neuroscience, 17, 1371145-1371145 (2024-04-04)
The prevailing model behind synapse development and specificity is that a multitude of adhesion molecules engage in transsynaptic interactions to induce pre- and postsynaptic assembly. How these extracellular interactions translate into intracellular signal transduction for synaptic assembly remains unclear. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service