Skip to Content
MilliporeSigma

ABF108

Anti-IL-33 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Interleukin-33, IL-33, Interleukin-33(102-266), Interleukin-33(109-266)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
FACS, WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

technique(s)

flow cytometry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Il33(77125)

General description

IL-33 is a proinflammatory protein that shares structural and functional characteristics with the IL-1 cytokine family. It binds and signals through the IL-1RL1/ST2 receptor, leading to the activation of MAP kinases and NF-kappaB. IL-33 induces production of helper T-cell type 2 (TH2) cytokines, including IL-4, IL-5 and IL-13, and is a known mediator of many inflammation responses. Cat. No. ABF108, Anti-IL-33, is an affinity purified rabbit polyclonal antibody that recognizes epitopes in the C-terminal half of murine IL-33. Reactivity to rat or other species has not been tested.
~35 kDa observed. Uncharacterized band(s) may appear in some lysates.

Immunogen

Epitope: C-Terminal
His-tagged recombinant protein corresponding to the C-Terminal of mouse IL-33.

Application

Anti-IL-33 Antibody is an antibody against IL-33 for use in Western Blotting, Flow Cytometry.
Flow Cytometry Analysis: 0.25 µg of this antibody from a representative lot detected IL-33 in 1X10E6 monensin treated MEF1 cells.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Physical form

Affinity
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in MEF1 cell lysate.

Western Blotting Analysis: 0.1 µg/mL of this antibody detected IL-33 in 10 µg of MEF1 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library