Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
1
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
technique(s)
immunohistochemistry: suitable (paraffin), immunoprecipitation (IP): suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... XRCC6(2547)
General description
Lupus Ku autoantigen protein p70 (Ku70) is also known as X-ray repair cross-complementing protein 6 (XRCC6), 5′-dRP lyase Ku70, CTC box-binding factor 75 kDa subunit (CTC75, CTCBF) or Thyroid-lupus autoantigen (TLAA). Ku70 is a single stranded DNA-dependent ATP-dependent helicase that plays a role in chromosome translocation and DNA non-homologous end joining (NHEJ) required for double-strand break repair and recombination. The XRCC5/6 (Ku80/Ku70) dimer acts as regulatory subunit of the DNA-dependent protein kinase complex DNA-PK, and that dimer, together with APEX1, acts as a negative regulator of transcription. Individuals with systemic lupus erythematosus (SLE) and related disorders produce extremely large amounts of autoantibodies to XRCC5 (Ku80) and XRCC6 (Ku70).
~70 kDa observed
Immunogen
Epitope: Near N-Terminus
KLH-conjugated linear peptide corresponding to human Ku70 Protein near N-Terminus.
Application
Detect Ku70 Protein using this rabbit polyclonal antibody, Anti-Ku70 Protein Antibody validated for use in western blotting, IP & IHC (Paraffin).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Ku70 Protein in 0.5 mg of HeLa cell lysate.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Ku70 Protein in human prostate carcinoma cells and human testis tissue.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Ku70 Protein in human prostate carcinoma cells and human testis tissue.
Research Category
Epigenetics & Nuclear Function
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
Chromatin Biology
Biochem/physiol Actions
This antibody recognizes Ku70 Protein near N-Terminus.
Physical form
Format: Purified
Protein A purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Control
HeLa cell lysate
HeLa cell lysate
Evaluated by Western Blotting in HeLa cell lysate.
Western Blotting Analysis: 0.5 µg/mL of this antibody detected Ku70 Protein in 10 µg of HeLa cell lysate.
Western Blotting Analysis: 0.5 µg/mL of this antibody detected Ku70 Protein in 10 µg of HeLa cell lysate.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: AB1358
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.