Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABE541

Sigma-Aldrich

Anti-phospho-STAT2 (Tyr689) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

p113

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr689)

Gene Information

human ... STAT2(6773)

General description

The Janus kinase (JAK)/signal transducers and activators of transcription (STAT) signaling pathway plays an important role in cell proliferation, cell differentiation, cell migration, and cell death. It is the principal signaling mechanism for a variety of cytokines and growth factors. Constitutive activation or dysregulation of JAK/STAT signaling can result in inflammatory disease, erythrocytosis, gigantism, and leukemia. STAT2 (113 kDa), originally purified from the nuclei of a-interferon-treated cells, is critical to the transcriptional responses induced by type I interferons, IFN-alpha/beta. Knockout mice with a targeted disruption of STAT2 exhibit greater susceptibility to viral infection and altered responses to Type I interferons. Stat2 is rapidly activated by phosphorylation at Tyr690 in response to stimulation by IFN-alpha/beta via assocations with receptor-bound Jak kinases.

Specificity

Other homologies: Rat (89% sequence homology).
This antibody recognizes STAT2 phosphorylated at Tyr689.

Immunogen

Epitope: Phosphorylated Tyr689.
KLH-conjugated linear peptide corresponding to mouse STAT2 phosphorylated at Tyr689.

Application

Anti-phospho-STAT2 (Tyr689) Antibody is an antibody against phospho-STAT2 (Tyr689) for use in Immunoprecipitation.
Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated phospho-STAT2 (Tyr689) from IFN alpha treated HeLa cell lysate.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Immunological Signaling

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: A 1:200 dilution of this antibody detected phospho-STAT2 (Tyr689) in 10 µg of IFN alpha/gamma treated HeLa cell lysate.

Target description

~113 kDa observed. An uncharacterized band may appear at ~260 kDa in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Untreated and IFN alpha/gamma treated HeLa cell lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service