Skip to Content
MilliporeSigma

ABE52

Anti-Cyclin D Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

B-cell CLL/lymphoma 1, BCL-1 oncogene, G1/S-specific cyclin D1, PRAD1 oncogene, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IP, WB
Citations:
7
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

primate (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CCND1(595)

General description

Progression through G1 phase and transition from G1 to S phase of the cell division cycle is controlled by activation of a distinct series of serine/threonine kinase complexes which comprise of a cyclin regulatory subunit and a cyclin dependent kinase (cdk). D type cyclins are induced earlier than cyclin E in G1 and can form complexes with cdk2,-4,-5, and -6. Cyclin D1 is a nuclear protein during G1 phase and disappears from the nucleus in S-phase. Among the key cell cycle regulators, cyclin D1 has been implicated most strongly as a proto-oncogene in several human tumor types, including breast carcinomas.
~ 34 kDa

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to human Cyclin D at the C-terminus.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Use Anti-Cyclin D Antibody (Rabbit Polyclonal Antibody) validated in WB, ICC, IP to detect Cyclin D also known as B-cell CLL/lymphoma 1, BCL-1 oncogene.
Western Blot (SNAP ID) Analysis: 5 µg/ml from a previous lot detected Cyclin D on 10 µg of 3T3/A31 cell lysate.

Immunocytochemistry Analysis: 1:500 dilution from a previous lot detected Cyclin D in NIH/3T3, A431, C2C12, and HeLa cells.

Biochem/physiol Actions

This antibody recognizes Cyclin D at the C-terminus.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
3T3/A31 cell lysate
Evaluated by Western Blot in 3T3/A31 cell lysate.

Western Blot Analysis: 1 µg/ml of this antibody detected Cyclin D on 10 µg of 3T3/A31 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 04-221

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Hsing-Yu Chen et al.
International journal of molecular sciences, 24(13) (2023-07-14)
Contrast-induced nephropathy (CIN) is one of the most common causes of acute kidney injury (AKI). However, management is still limited, and the cellular response to radiocontrast removal for CIN remains unclear. This study aimed to explore the latent effects of
Divya Gandra et al.
Cancer medicine, 13(9), e7207-e7207 (2024-04-30)
Most high-risk neuroblastoma patients who relapse succumb to disease despite the existing therapy. We recently reported increased event-free and overall survival in neuroblastoma patients receiving difluoromethylornithine (DFMO) during maintenance therapy. The effect of DFMO on cellular processes associated with neuroblastoma
Liping Ou et al.
Molecular medicine reports, 13(1), 720-730 (2015-12-10)
Embryonic stem cells (ESCs) are pluripotent stem cells derived from early stage embryos. It remains unclear whether inhibiting the Wnt/β‑catenin signaling pathway using dickkopf Wnt signaling pathway inhibitor 1 (DKK1) impacts on the differentiation potential of mouse ESCs in vitro



Global Trade Item Number

SKUGTIN
ABE5204053252512896