Skip to Content
MilliporeSigma

ABE365

Anti-LSD1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Lysine-specific histone demethylase 1A, BRAF35-HDAC complex protein BHC110, Flavin-containing amine oxidase domain-containing protein 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KDM1A(23028)

General description

LSD1 (lysine specific demethylase 1), also known as BHC110, is a histone demethylase that specifically demethylates Lys4 of histone H3, a specific tag for epigenetic transcriptional activation, thereby acting as a corepressor. LSD1 acts by oxidizing the substrate by FAD to generate the corresponding imine that is subsequently hydrolyzed. LSD1 demethylates both mono- and tri-methylted Lys4 of histone H3, and may play a role in the repression of neuronal genes. Alone, it is unable to demethylate H3 Lys4 on nucleosomes and requires the presence of RCOR1/CoREST to achieve such activity. LSD1 may also demethylate Lys9 of histone H3, a specific tag for epigenetic transcriptional repression, thereby leading to derepression of androgen receptor target genes.
~110 kDa observed. Uniprot describes two isoforms at ~92 kDa and ~95 kDa due to alternative splicing.

Immunogen

Histidine-tagged recombinant protein corresponding to full length LSD1.

Application

Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated LSD1 in 0.5 mg of HeLa nuclear extract.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-LSD1 Antibody (Rabbit Polyclonal Antibody) validated in WB, IP to detect LSD1 also known as Lysine-specific histone demethylase 1A, BRAF35-HDAC complex protein BHC110.

Biochem/physiol Actions

This antibody recognizes LSD1.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa nuclear extract
Evaluated by Western Blot in HeLa nuclear extract.

Western Blot Analysis: 0.05 µg/mL of this antibody detected LSD1 in 10 µg of HeLa nuclear extract.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: MABE545

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library