Skip to Content
MilliporeSigma

ABE244

Anti-monomethyl-Histone H3 Antibody (Lys36)

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H3 (monomethyl Lys36), H3K36me, H3 histone family, member T, Histone 3, H3, Histone cluster 3, H3

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
mouse, human, rat
Application:
DB, IP, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

technique(s)

dot blot: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

monomethylation (Lys36)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is one of four histones that form a core octameric structure that is required for the packaging of DNA into nucleosomes, the most basic level of chromatin arrangement. Histones undergo a number of post-translational modifications (PTM) at their N- and C-terminal tails which can alter chromatin structure and stability, and influence DNA repair and gene expression. Histone H3 undergoes methylation of its lysine and arginine residues, with varying biological outcomes. The lysine 36 residue undergoes mono-, di-, or trimethylation which may be performed by one of many methyltransferase enzymes (SET2, NSD1, NSD2, NSD3, ASH1, SMYD2, SETMAR, KMT3A, and SETD3). Several studies have indicated that methylated lysine 36 residues promote Rpd3S-mediated deacetylation of other histones resulting in gene activation.
~17 kDa observed

Immunogen

Epitope: Monomethylated Lys36
KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H3 monomethylated at Lys36.

Application

Anti-monomethyl-Histone H3 (Lys36) Antibody is a rabbit polyclonal antibody for detection of Histone H3 monomethylated at lysine 36. This purified polyclonal antibody also known as Anti-H3K36me1, has dot blot (DB) proven specificity & has been validated in WB, IP.
Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated monomethyl-Histone H3 (Lys36) from HeLa acid extract.

Dot Blot Specificity Analysis: Unmodified and various modified Histone peptides (see table) were probed with Anti-monomethyl-Histone H3 (Lys36) (0.5 µg/mL).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Biochem/physiol Actions

This antibody recognizes the N-terminus of Histone H3 monomethylated at Lys36.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa acid extract and various recombinant Histone proteins
Evaluated by Western Blot in HeLa acid extract and various recombinant Histone proteins.

Western Blot Analysis: 0.05 µg/mL of this antibody detected monomethyl-Histone H3 (Lys36) in 10 µg of HeLa acid extract.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Cancer is a complex disease manifestation. At its core, it remains a disease of abnormal cellular proliferation and inappropriate gene expression. In the early days, carcinogenesis was viewed simply as resulting from a collection of genetic mutations that altered the gene expression of key oncogenic genes or tumor suppressor genes leading to uncontrolled growth and disease (Virani, S et al 2012). Today, however, research is showing that carcinogenesis results from the successive accumulation of heritable genetic and epigenetic changes. Moreover, the success in how we predict, treat and overcome cancer will likely involve not only understanding the consequences of direct genetic changes that can cause cancer, but also how the epigenetic and environmental changes cause cancer (Johnson C et al 2015; Waldmann T et al 2013). Epigenetics is the study of heritable gene expression as it relates to changes in DNA structure that are not tied to changes in DNA sequence but, instead, are tied to how the nucleic acid material is read or processed via the myriad of protein-protein, protein-nucleic acid, and nucleic acid-nucleic acid interactions that ultimately manifest themselves into a specific expression phenotype (Ngai SC et al 2012, Johnson C et al 2015). This review will discuss some of the principal aspects of epigenetic research and how they relate to our current understanding of carcinogenesis. Because epigenetics affects phenotype and changes in epigenetics are thought to be key to environmental adaptability and thus may in fact be reversed or manipulated, understanding the integration of experimental and epidemiologic science surrounding cancer and its many manifestations should lead to more effective cancer prognostics as well as treatments (Virani S et al 2012).

Global Trade Item Number

SKUGTIN
ABE24404053252551192

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service