Skip to Content
MilliporeSigma

ABE24

Anti-BPTF Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Protein Bptf, Bromodomain PHD-finger Transcription Factor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
3
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BPTF(2186)
mouse ... Bptf(207165)

General description

Bromodomain PHD finger transcription factor (UniProt: E9Q6A7; also known as Nucleosome-remodeling factor subunit BPTF; Fetal Alz-50 clone 1 protein, Fetal Alzheimer antigen) is encoded by the Bptf (also known as Fac1, Falz) gene (Gene ID: 207165) in murine species. BPTF, SNF2L and pRBAP46/48 are the three subunits of the nucleosome-remodelling factor NURF, an ISWI chromatin-remodeling complex that catalyzes ATP-dependent nucleosome sliding. BPTF mediates NURF target site specificity via interactions with transcription factors, histone variants and histone modifications on transcriptionally active genes. BPTF plays an important role in regulating nucleosome occupancy at nucleosome-free regions (NFRs) at sites occupied by the multivalent factors CTCF and cohesin via direct interaction with CTCF and the cohesin subunit SA2. In addition, BPTF also interacts with c-MYC and plays an essential role in its genomic distribution and biological activity. BPTF is necessary for the survival of c-MYC-overexpressing cells and for c-MYC-driven tumorigenesis in the mouse pancreas. BPTF knockdown leads to decreased c-MYC chromatin accessibility and significantly delays tumour development in pre-neoplastic pancreatic acinar cells. Consistently, BPTF expression in human tumours positively correlates with activation of c-MYC gene signatures.
Target molecular weight ~250 kDa observed. 321.6 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

GST-tagged recombinant N-terminal fragment of mouse BPTF.

Application

Anti-BPTF, Cat. No. ABE24, is a rabbit polyclonal antibody that detects BPTF and is tested for use in Western Blotting, Immunocytochemistry, Immunoprecipitation, and Peptide Inhibition Assay.
Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected BPTF in NIH/3T3 cell lysate.

Tested applications

Western Blotting Analysis: A 1:2,000 dilution from a representative lot detected BPTF in MEF-1 and mESC cell lysates.

Immunoprecipitation: A representative lot immunoprecipitated BPTF in Immunoprecipitation application (Qiu, Z., et al. (2015). Mol. Cell. Biol. 35(1):224-237).

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected BPTF in A431 cells.

Peptide Inhibition Assay: Target band detection in lysate from NIH/3T3 cells was prevented by pre-blocking of a representative lot with recombinant fragment of mouse BPTF.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Research Category
Epigenetics & Nuclear Function

Biochem/physiol Actions

This rabbit polyclonal antibody detects BPTF. It targets an epitope within 103 amino acids from the N-terminal half.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library