Skip to Content
MilliporeSigma

ABE21

Anti-Smad4/DPC4 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Mothers against decapentaplegic homolog 4, MAD homolog 4, Mothers against DPP homolog 4, Deletion target in pancreatic carcinoma 4, SMAD family member 4, SMAD 4, Smad4, hSMAD4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SMAD4(4089)

General description

SMAD4 is a member of the Mothers Against Dpp (MAD)-related family of proteins. SMAD4 is functionally distinct among the SMAD family, and is required for the assembly and transcriptional activation of diverse, SMAD-DNA complexes. SMAD4 appears to be not regulated by phosphorylation, but acts as a common mediator of TGF-β, activin, and bone morphogenetic protein signaling responses. SMAD4 is frequently inactivated in pancreatic, biliary and colorectal tumors.
~68 kDa observed.

Immunogen

Epitope: SAD region
GST-tagged recombinant protein corresponding to human Smad4 at the SAD region.

Application

Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected Smad4 in NIH/3T3, HeLa, and A431 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-Smad4/DPC4 Antibody (Rabbit Polyclonal Antibody) validated in WB, ICC to detect Smad4/DPC4 also known as Mothers against decapentaplegic homolog 4, SMAD 4, Smad4, hSMAD4.

Biochem/physiol Actions

This antibody recognizes Smad4 at the SAD region.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HepG2 cell lysate
Evaluated by Western Blot in HepG2 cell lysate.

Western Blot Analysis: 0.1 µg/mL of this antibody detected Smad4 on 10 µg of HepG2 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 04-1033

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library