Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE1971-I

Sigma-Aldrich

Anti-phospho TFEB (Ser142)

from rabbit, purified by affinity chromatography

Synonym(s):

Transcription factor EB, Class E basic helix-loop-helix protein 35, bHLHe35

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

phosphorylation (pSer142)

Gene Information

human ... TFEB(7942)

General description

Transcription factor EB (UniProt: P19484; also known as Class E basic helix-loop-helix protein 35, bHLHe35) is encoded by the TFEB (also known as BHLHE35) gene (Gene ID: 7942) in human. TFEB is a basic Helix-Loop-Helix (bHLH) leucine zipper transcription factor that plays a major role in the regulation of lysosomal biogenesis. TFEB is also a main player in the transcriptional response to starvation and controls autophagy by positively regulating autophagosome formation and autophagosome-lysosome fusion. TFEB is phosphorylated by mTORC1 and ERK at Ser142, which prevents its nuclear translocation. Pharmacological inhibition of mTORC1, as well as starvation and lysosomal disruption, activates TFEB nuclear translocation. Similarly, the serine-threonine kinase RIP1/RIPK1 is shown to suppress basal autophagic flux by activating ERK, which in turn phosphorylates TFEB at Ser142 and thereby prevents TFEB-mediated transcription of gene products involved in autophagy.

Specificity

This rabbit polyclonal antibody specifically detects Transcription factor EB phosphorylated on Serine 142 in human cells.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids surrounding phosphoserine 142 from the N-terminal half of human Transcription factor EB, isoform 1.

Application

Anti-phospho TFEB (Ser142), Cat. No. ABE1971-I, is a highly specific rabbit polyclonal antibody that targets Transcription factor EB phosphorylated on Serine 142 and has been tested for use in Western Blotting.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in transfected HEK293 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected phospho TFEB (Ser142) in lysate of HEK293 cells transfected with FLAG-tagged wild type TFEB.

Target description

~65 kDa observed; 52.87 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuzuki Kano et al.
Kidney360, 5(2), 182-194 (2023-12-08)
Collectrin serves as a chaperone for the trafficking of neutral amino acid (AA) transporters in the apical membranes of proximal tubular cells (PTCs). Cltrn knockout reduced AAs influx into PTCs, inactivated mTOR, activated transcription factor EB, improved lysosome function, and
Zhi-Hang Li et al.
Journal of cellular and molecular medicine, 25(12), 5729-5743 (2021-05-06)
Cyclosporine A (CsA) is an immunosuppressor widely used for the prevention of acute rejection during solid organ transplantation. However, severe nephrotoxicity has substantially limited its long-term usage. Recently, an impaired autophagy pathway was suggested to be involved in the pathogenesis
Jin-Feng Zhao et al.
iScience, 27(8), 110432-110432 (2024-07-31)
Reversible phosphorylation of the transcription factor EB (TFEB) coordinates cellular responses to metabolic and other stresses. During nutrient replete and stressor-free conditions, phosphorylated TFEB is primarily localized to the cytoplasm. Stressor-mediated reduction of TFEB phosphorylation promotes its nuclear translocation and
Lizhen Cheng et al.
Cell communication and signaling : CCS, 21(1), 91-91 (2023-05-05)
Diabetic encephalopathy (DE) is a complication of type 2 diabetes mellitus (T2DM) that features Alzheimer's disease (AD)-like pathology, which can be degraded by the autophagy-lysosome pathway (ALP). Since transcription factor EB (TFEB) is a master regulator of ALP, TFEB-mediated ALP
Maurizio Forte et al.
Autophagy, 19(4), 1087-1099 (2022-08-24)
NPPA/atrial natriuretic peptide (natriuretic peptide type A) exerts critical pleiotropic effects in the cardiovascular system, limiting cardiomyocyte hypertrophy and death, reducing cardiac fibrosis and promoting vascular integrity. However, the molecular mechanisms underlying these beneficial effects still need to be clarified.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service