Skip to Content
MilliporeSigma
All Photos(1)

Documents

ABE1863

Sigma-Aldrich

Anti-PAGR1 (PA1)

from rabbit

Synonym(s):

PAXIP1-associated glutamate-rich protein 1, Glutamate-rich co-activator interacting with SRC11, GAS1, PAXIP1-associated protein 1, PTIP-associated protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

ChIP: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PAGR1(79447)

General description

PAXIP1-associated glutamate-rich protein 1 (UniProt: Q9BTK6; also known as Glutamate-rich co-activator interacting with SRC1, GAS, PAXIP1-associated protein 1, PTIP-associated protein 1) is encoded by the PAGR1 (also known as C16orf53, PA1) gene (Gene ID: 79447) in human. It associates with the histone methyltransferase MLL3/4 complex, which suggests that it plays a role in epigenetic and transcriptional activation. However, in association with PAXIP1/PTIP is proposed to function at least in part independently of the MLL3/4 complex. It is proposed to be recruited by PAXIP1 to sites of DNA damage where the PAGR1:PAXIP1 complex is required for cell survival in response to DNA damage independently of the MLL3/4 complex. During immunoglobulin class switching in activated B-cells, it is involved in transcription regulation of downstream switch regions at the immunoglobulin heavy-chain (Igh) locus independently of the MLL3/4 complex. PAGR1 protein is reported to act as a transcriptional cofactor for estrogen receptor. It interacts with GR (NR3C1) and inhibits the GR transactivation.

Specificity

This polyclonal antibody detects PAXIP1-associated glutamate-rich protein 1 in human and mouse.

Immunogen

His-tagged full-length human recombinant PAXIP1-associated protein 1.

Application

Anti-PAGR1 (PA1), Cat. No. ABE1863, is a highly specific rabbit polyclonal antibody that targets PAXIP1-associated glutamate-rich protein 1 and has been tested in Chromatin Immunoprecipitation (ChIP), Immunocytochemistry, and Western Blotting.
Western Blotting Analysis: A representative lot detected PAGR1 (PA1) in Western Blotting applications (Lee, J.E., et. al. (2013). Elife. 2:e01503; Gong, Z., et. al. (2009). J Biol Chem. 284(11):7284-93).

Chromatin Immunoprecipitation Analysis: A representative lot detected PAGR1 (PA1) in MEF′s (Cho, Y.W., et. al. (2009). Cell Metab. 10(1):27-39).

Immunocytochemistry Analysis: A representative lot detected PAGR1 (PA1) in U2OS & HeLa cells (Cho, Y.W., et. al. (2009). Cell Metab. 10(1):27-39).

Quality

Evaluated by Western Blotting in HEK293 cell nuclear extract.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected PAGR1 (PA1) in 10 µg of HEK293 cell nuclear extract.

Target description

~42 kDa observed; 27.72 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service