Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE1391

Sigma-Aldrich

Anti-Brd4 Antibody

serum, from rabbit

Synonym(s):

Bromodomain-containing protein 4, Mitotic chromosome-associated protein, MCAP, Brd4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

mouse

technique(s)

ChIP: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Brd4(57261)

General description

Bromodomain-containing protein 4 (Brd4), or Mitotic chromosome-associated protein (MCAP), is a chromatin reader protein and kinase that recognizes and binds acetylated histones and plays a key role in the transmission of epigenetic memory across cell divisions and transcription regulation. Brd4 remains associated with acetylated chromatin throughout the entire cell cycle and provides epigenetic memory for post mitotic G1 gene transcription by preserving acetylated chromatin status and maintaining high-order chromatin structure. During interphase, Brd4 plays a key role in regulating the transcription of signal-inducible genes by associating with the P-TEFb complex and recruiting it to promoters. Brd4 is widely expressed and localized to the nucleus and acetylated chromatin. Brd4 also plays a large role in many cancers, and through its regulation of transcription, Brd4 is important in multiple cellular processes including DNA damage responses, aging, and development.

Specificity

Recognizes human Brd4

Immunogen

Epitope: Region between amino acids 150 to 290 of human Brd4
Recombinant protein corresponding to Mouse Brd4 short form.

Application

Research Category
Epigenetics & Nuclear Function

Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Histones
This Anti-Brd4 Antibody is validated for use in Western Blotting and Chromatin Immunoprecipitation (ChIP) and Immunocytochemistry for the detection of Brd4.
Western Blotting Analysis: A representative lot of this antibody was reported to detect Brd4 protein in a HeLa cell lysate (Alsarraj J., et al. (2013). Also see Anup, D., et al. (2003) and Mochizuki, K., et al. (2008) for additional data using mouse embyronic fibroblasts and NIH3T3 cells.

Chromatin Immunoprecipitation (ChIP): A representative lot of this antibody was reported to work in ChIP using chromatin from NIH3T3 cells. See Mochizuki, K., et al. (2008) and Anup, D., et al. (2009).

Immunocytochemistry: A representative lot of this antibody was reported to work in ICC. See Anup, D., et al. (2000) and Anup, D., et al. (2009).

Quality

Evaluated by Western Blotting in HEK293 cells.

Western Blotting Analysis: 1:1,000 dilution of this antibody detected Brd4 in 10 µg of HEK293 cell lysate.

Target description

~ 100 kDa observed
This antibody primarily detects 100 kd short form of Brd4 and to lesser extent larger 180 kD form (see Alsarraj J., et al. (2013 for additional details on performance of this antibody).

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Talha Ijaz et al.
Cell death & disease, 8(2), e2606-e2606 (2017-02-10)
Transdifferentiation of quiescent dermal fibroblasts to secretory myofibroblasts has a central role in wound healing and pathological scar formation. This myofibroblast transdifferentiation process involves TGFβ-induced de novo synthesis of alpha smooth muscle cell actin (αSMA)+ fibers that enhance contractility as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service