Skip to Content
MilliporeSigma

ABD38

Anti-SNAI1 (Snail homolog 1) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Zinc finger protein SNAI1, Protein snail homolog 1, Protein sna

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SNAI1(6615)

General description

SNAI1, also known as Snail homolog 1, is a transcription factor that down regulates the expression of ectodermal genes within the mesoderm and is thought to be essential for mesoderm formation in the developing embryo. SNAI1 is expressed in the kidney and in mesenchymal and epithelial cell lines. Mutations in SNAI1 have been associated with tumors.
~29 kDa observed. An uncharacterized band appears at ~40 kDa in some lysates.

Immunogen

Linear peptide corresponding to human SNAI1.

Application

Anti-SNAI1 (Snail homolog 1) Antibody is a Rabbit Polyclonal Antibody for detection of SNAI1 also known as Zinc finger protein SNAI1, Snail homolog 1 & has been validated in WB.
Research Category
Stem Cell Research
Research Sub Category
Transcription Factors
Western Blot Analysis: 1 µg/mL from a representative lot detected SNAI1 on 10 µg of mouse brain tissue lysate.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain tissue lysate
Evaluated by Western Blot in rat brain tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected SNAI1 on 10 µg of rat brain tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library