Skip to Content
MilliporeSigma

ABD12

Anti-Aldehyde dehydrogenase 1 (ALDH1A1) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

aldehyde dehydrogenase 1 family, member A1, ALDH class 1, Aldehyde dehydrogenase, cytosolic, retinaldehyde dehydrogenase 1, Aldehyde dehydrogenase family 1 member A1, retinal dehydrogenase 1, acetaldehyde dehydrogenase 1, aldehyde dehydrogenase 1, solubl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ALDH1A1(216)

General description

Aldehyde dehydrogenase 1A1 (ALDH1A1) is also known as retinal dehydrogenase 1. It is thought to be a main converter of retinol to retinoic acid, which is very important in cell to cell signaling. ALDH1A1 may also be involved in inhibiting invasion in pancreatic cancer cells. More specifically, ALDH1A1 may regulate hematopoietic stem cell differentiation by disrupting the retinoid signaling.
~54 kDa observed

Immunogen

KLH-conjugated linear peptide corresponding to human Aldehyde dehydrogenase 1.

Application

Research Category
Stem Cell Research
This Anti-Aldehyde dehydrogenase 1 (ALDH1A1) Antibody is validated for use in WB for the detection of Aldehyde dehydrogenase 1 (ALDH1A1).

Biochem/physiol Actions

Other homologies: rat and mouse (91% sequence homology).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HepG2 cell lysate
Evaluated by Western Blot in HepG2 cell lysate.

Western Blot Analysis: 0.05 µg/mL of this antibody detected Aldehyde dehydrogenase 1 on 10 µg of HepG2 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Natalia Ninkina et al.
Neurobiology of aging, 91, 76-87 (2020-04-01)
The etiology and pathogenesis of Parkinson's disease (PD) are tightly linked to the gain-of-function of α-synuclein. However, gradual accumulation of α-synuclein aggregates in dopaminergic neurons of substantia nigra pars compacta (SNpc) leads to the depletion of the functional pool of
Yi Sui et al.
British journal of cancer, 125(7), 1003-1015 (2021-07-12)
Metastasis is the major cause of treatment failure and cancer-related deaths in prostate cancer (PCa) patients. Our previous study demonstrated that a CD44+ subpopulation isolated from PCa cells or tumours possesses both stem cell properties and metastatic potential, serving as
Yi Sui et al.
Annals of translational medicine, 9(1), 45-45 (2021-02-09)
Prostate cancer (PCa) is one of the most common types of cancer and the emerging resistance to androgen deprivation therapy in PCa aggravates disease progression. In this study, we examined the potential pro-tumorigenic functions of NANOGP8 in prostate cancer development.



Global Trade Item Number

SKUGTIN
ABD12-25UG04054839337376