Skip to Content
MilliporeSigma

ABC41-M

Anti-Mitofusin-1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Fzo homolog, Transmembrane GTPase MFN1, Mitofusion-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat, mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... MFN1(55669)

General description

Along with Mitofusin-2 (Mfn2), Mitofusin-1 (Mfn1) is a known protein mediator of mitochondrial fusion. Mitochondrial fusion regulates mitochondrial morphogensis and supports complementation between mitochondrial genomes in mammalian cells. Two Mfn1 transcripts are detected at similar levels in a variety of human tissues and are dramatically elevated in the heart. Human Mfn1 is known to be localized in the mitochondria. It is reported that point mutations in the conserved G1 and G2 region of the Mfn1 GTPase domain result in decreased formation and fragmentation of the mitochondria, respectively.
~86 kDa observed.
Three isoforms of Mitofusin-1 are produced by alternative splicing with molecular weights of ~42 kDa, ~71 kDa, and ~84 kDa Uncharacterized bands may appear at ~45 kDa, ~48 kDa, and ~55 kDa in some lysates.

Immunogen

Epitope: Cytoplasmic Domain
KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human Mitofusin-1.

Application

Detect Mitofusin-1 using this Anti-Mitofusin-1 Antibody validated for use in WB.
Research Category
Cell Structure
Research Sub Category
Organelle & Cell Markers

Biochem/physiol Actions

This antibody recognizes Mitofusin-1 at the cytoplasmic domain.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate
Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Mitofusin-1 in 10 µg HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library