Skip to Content
MilliporeSigma

Skip To

AB9916

Anti-CYP2b10 Antibody

serum, Chemicon®

Synonym(s):

Cytochrome P450 2b10

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
11
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Cyp2B10(13088)

General description

58kDa
CYP2b10 (Cytochrome P450, family 2, subfamily b, polypeptide 10) is a member of the cytochrome P450 superfamily of monooxygenase enzymes which catalyze many reactions involved in drug metabolism and the synthesis of cholesterol, steroids and other lipids.

Immunogen

Epitope: AA I182 – T205
Recombinant 2b10

Application

This Anti-CYP2b10 Antibody is validated for use in WB for the detection of CYP2b10.

Biochem/physiol Actions

AB9916 is specific for CYP2b10.

Analysis Note

Routinely evaluated by immunoblot.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB10320AB1252MABS1251
description

serum, Chemicon®

description

serum, Chemicon®

description

serum, Chemicon®

description

clone 41-17B, from mouse

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

serum

antibody form

serum

antibody form

serum

antibody form

purified immunoglobulin

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

mouse

technique(s)

western blot: suitable

technique(s)

western blot: suitable

technique(s)

western blot: suitable, immunohistochemistry: suitable

technique(s)

western blot: suitable, immunohistochemistry: suitable, immunofluorescence: suitable

UniProt accession no.

P12791

UniProt accession no.

Q16678

UniProt accession no.

P05181

UniProt accession no.

P19099


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters