Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB9608

Sigma-Aldrich

Anti-Neuropeptide Y Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-PYY4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NPY(4852)

Specificity

Neuropeptide Y.

Immunogen

Synthetic Neuropeptide Y.

Application

Immunohistochemistry: 1:500-1:1,000 on formalin-fixed, paraffin embedded human brain sections and rat brain and spinal cord. It is necessary to pretreat the section with heat induced epitope retrieval. EDTA, pH 8.0 is recommended.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Hormones & Receptors
This Anti-Neuropeptide Y Antibody is validated for use in IH for the detection of Neuropeptide Y.

Linkage

Replaces: AB1915

Physical form

Format: Purified
Purified immunoglobulin in PBS with 0.1% sodium azide. Liquid.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sahithi Attaluri et al.
Frontiers in aging neuroscience, 15, 1200445-1200445 (2023-07-10)
Extracellular vesicles (EVs) released by human-induced pluripotent stem cell (hiPSC)-derived neural stem cells (NSCs) have robust antiinflammatory and neurogenic properties due to therapeutic miRNAs and proteins in their cargo. Hence, hiPSC-NSC-EVs are potentially an excellent biologic for treating neurodegenerative disorders
Uthra Rajamani et al.
Cell stem cell, 22(5), 698-712 (2018-04-24)
The hypothalamus contains neurons that integrate hunger and satiety endocrine signals from the periphery and are implicated in the pathophysiology of obesity. The limited availability of human hypothalamic neurons hampers our understanding of obesity disease mechanisms. To address this, we
Wei-Kai Huang et al.
Cell stem cell, 28(9), 1657-1670 (2021-05-08)
Human brain organoids represent remarkable platforms for recapitulating features of human brain development and diseases. Existing organoid models do not resolve fine brain subregions, such as different nuclei in the hypothalamus. We report the generation of arcuate organoids (ARCOs) from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service