Skip to Content
MilliporeSigma

AB9415

Anti-Cannabinoid Receptor 1 Antibody

Chemicon®, from rabbit

Synonym(s):

CB1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IHC
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

monkey, human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable (paraffin)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CNR1(1268)

Immunogen

Synthetic peptide from the 3rd cytoplasmic domain of human Cannabinoid Receptor 1. The immunogen sequence has 100% homology with mouse.

Application

Anti-Cannabinoid Receptor 1 Antibody detects level of Cannabinoid Receptor 1 & has been published & validated for use in IC, IH(P).
Immunohistochemistry on formalin fixed paraffin embedded tissue sections.

Immunocytochemistry on acetone fixed cells.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neuroinflammation & Pain

Neurotransmitters & Receptors

Biochem/physiol Actions

Recognizes Cannabinoid Receptor 1 [CB1].

Physical form

Affinity purified immunoglobulin. Liquid in PBS, with less than 0.1% sodium azide.

Preparation Note

Maintain at -70°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library