Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB9282

Sigma-Aldrich

Anti-Peripherin Antibody

Chemicon®, from chicken

Synonym(s):

Anti-NEF4, Anti-PRPH1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

chicken

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Specificity

Peripherin.

Immunogen

Recombinant full length peripherin.

Application

Detect Peripherin using this Anti-Peripherin Antibody validated for use in WB & IC.
Research Category
Neuroscience
Research Sub Category
Sensory & PNS

Neuronal & Glial Markers
Western blot: 1:5,000-1:10,000. The antibody reacts with the ~57 kDa peripherin protein.

Immunocytochemistry: 1:500-1:1,000

Optimal working dilutions must be determined by the end user.

Physical form

Chicken IgY. Liquid containing ~10 mM sodium azide.
Format: Purified

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
POSITIVE CONTROL: Rat spinal cord or peripheral nerve homogenate.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dvir Blivis et al.
Scientific reports, 9(1), 3201-3201 (2019-03-03)
We investigated dye-coupling between motoneurons in the L6 segment of the neonatal mouse spinal cord that contains limb-innervating motoneurons and sexually dimorphic motor nuclei. Using an isolated spinal cord preparation, we back-filled the cut, L6 ventral root with the small
Nancy E J Berman et al.
Headache, 46(8), 1230-1245 (2006-09-01)
We examined changes in the serotonin system across the estrous cycle in trigeminal ganglia of female rodents to determine which components are present and which are regulated by the variations in levels of ovarian steroids that occur during the estrous
Deepthi Thomas et al.
International journal of physiology, pathophysiology and pharmacology, 12(6), 153-165 (2021-01-28)
Electrical synapses formed by gap junctions occur at a variety of neuronal subcellular sites in the mammalian central nervous system (CNS), including at somatic, dendritic and axon terminal compartments. Numerous electrophysiological studies using mice and rats, as well as computer
Marián Haburčák et al.
Frontiers in synaptic neuroscience, 14, 995474-995474 (2022-10-18)
The Spontaneously Hypertensive Rat (SHR) has increased sympathetic drive to the periphery that precedes and contributes to the development of high blood pressure, making it a useful model for the study of neurogenic hypertension. Comparisons to the normotensive Wistar Kyoto
W Thomas Clarke et al.
Journal of cell science, 123(Pt 15), 2543-2552 (2010-07-01)
Syncoilin is an atypical type III intermediate filament (IF) protein, which is expressed in muscle and is associated with the dystrophin-associated protein complex. Here, we show that syncoilin is expressed in both the central and peripheral nervous systems. Isoform Sync1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service