Skip to Content
MilliporeSigma

AB9042

Anti-Brain Derived Neurotrophic Factor Antibody, pro

Chemicon®, from chicken

Synonym(s):

proBDNF, pro BDNF

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
9
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

chicken

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... BDNF(627)

Immunogen

Fusion protein from mouse proBDNF containing only the prodomain region.

Application

Anti-Brain Derived Neurotrophic Factor Antibody, pro detects level of Brain Derived Neurotrophic Factor & has been published & validated for use in IP, WB, IH.
Western blot: 1:500-1:1,000. Reacts with bands of 30 kD and 28 kD.Immunohistochemistry: 1:100-1:250 on frozen sections.

Immunohistochemistry: 1:100-1:200

Immunoprecipitation: 1:100

Optimal working dilutions must be determined by the end user.

Biochem/physiol Actions

Reacts with proBDNF. Does not recognize mature BDNF isoforms.

Physical form

Chicken immunoglobulin. Liquid.
Format: Semi-Purified

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library