Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB761

Sigma-Aldrich

Anti-Collagen Type II Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-Anti-ANFH, Anti-Anti-AOM, Anti-Anti-COL11A3, Anti-Anti-SEDC, Anti-Anti-STL1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... COL2A1(1280)

Specificity

The antibody reacts with both native and denatured human collagen type II.



Cross Reactivity Percent

Human collagen type II 90%

Human collagen type I, III, IV, V <1%

Human plasma proteins do not interfere with binding to collagen.

Immunogen

Collagen type II extracted and purified from human cartilage.

Application

Anti-Collagen Type II Antibody detects level of Collagen Type II & has been published & validated for use in ELISA, WB, IC, IH.
ELISA 1:1000.

Indirect immunofluorescence on cryostat sections or cultured cells 1:10-1:20

Western Blot 1:100

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Physical form

Rabbit antiserum passed over agarose sorbents with immobilized human blood plasma proteins and human collagen types I, III, IV and V. Antibody was affinity purified on immobilized human collagen type II.

Product is liquid in 0.15M sodium chloride, 10mM sodium phosphate pH 7.5 with 0.1% mannitol and 0.1% dextran as stabilizers. No preservative.

Storage and Stability

Maintain at 2-8°C for up to 12 months from date of reciept. Remove any nondispersed aggregates by microcentrifugation prior to use.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriella Nilsson Hall et al.
Biomaterials, 273, 120820-120820 (2021-04-20)
Tissue engineered constructs have the potential to respond to the unmet medical need of treating deep osteochondral defects. However, current tissue engineering strategies struggle in the attempt to create patterned constructs with biologically distinct functionality. In this work, a developmentally-inspired
Mu-En Lin et al.
Cardiovascular research, 112(2), 606-616 (2016-09-28)
Vascular smooth muscle cells (SMCs) are major precursors contributing to osteochondrogenesis and calcification in atherosclerosis. Runt-related transcription factor-2 (Runx2) has been found essential for SMC differentiation to an osteochondrogenic phenotype and subsequent calcification in vitro. A recent study using a
Fu-Hui Wang et al.
Scientific reports, 12(1), 21663-21663 (2022-12-16)
The human type II collagen (Col II), specifically expressed in chondrocytes, is a crucial component of the adult hyaline cartilage. We examine the potential of artificial induction of Col II in human peripheral blood mononuclear cells (PBMNCs) as a novel
Hannah Agten et al.
Frontiers in bioengineering and biotechnology, 12, 1386692-1386692 (2024-04-26)
Osteochondral defects are deep joint surface lesions that affect the articular cartilage and the underlying subchondral bone. In the current study, a tissue engineering approach encompassing individual cells encapsulated in a biocompatible hydrogel is explored in vitro and in vivo.
Gabriella Nilsson Hall et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 7(2), 1902295-1902295 (2020-01-30)
Clinical translation of cell-based products is hampered by their limited predictive in vivo performance. To overcome this hurdle, engineering strategies advocate to fabricate tissue products through processes that mimic development and regeneration, a strategy applicable for the healing of large

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service