Skip to Content
MilliporeSigma

AB6016

Anti-F-actin-capping protein subunit alpha-1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

capping protein (actin filament) muscle Z-line, alpha 1, CapZ alpha-1, F-actin-capping protein subunit alpha-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

pig, mouse, human, rat

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology), bovine (based on 100% sequence homology), primate (based on 100% sequence homology), canine (based on 100% sequence homology), porcine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable, western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

bovine ... Capza1(534656)
dog ... Capza1(475857)
human ... CAPZA1(829)
mouse ... Capza1(12340)
pig ... Capza1(100037957)
rat ... Capza1(691149)
rhesus monkey ... Capza1(705653)

General description

F-actin capping protein subunit alpha (CapZ alpha) belongs to the F-actin capping protein family, which are characteristic heterodimers, consisting of an α subunit (31-36 kDa) and a β subunit (28-32 kDa), that cap the barbed end of actin filaments within all eukaryotes. Their ability to bind the actin filaments is in a manner independent of Ca2++ and requires the C-terminal end of both subunits for optimal binding. CapZ is contained within the Z-discs of striated muscle, where it functions to inhibit the polymerization and depolymerization of actin. Within vertebrates, there are three isoforms for each of CapZ’s subunits. All three alpha subunit isoforms are components of the WASH complex. Little is known about isoforms 1 and 2; however, isoform 3 is thought to be involved in spermatid morphogenesis and is exclusively observed within sperm and testis, with highest expression within the neck region of sperm.
~35 kDa

Immunogen

GST-tagged recombinant protein corres-ponding to human F-actin-capping protein subunit alpha-1.

Application

Detect F-actin-capping protein subunit alpha-1 using this Anti-F-actin-capping protein subunit alpha-1 Antibody validated for use in WB & IC.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton
Western Blot (Snap i.d.) Analysis: 1 µg/mL from a previous lot detected F-actin-capping protein subunit alpha-1 on 10 µg of HeLa cell lysate.
Immunocytochemistry Analysis: A 1:500 dilution from a previous lot detected F-actin-capping protein subunit alpha-1 in NIH/3T3, HeLa, and A431 cells.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate
Evaluated by Western Blot in HeLa cell lysate.
Western Blot Analysis: 0.1 µg/mL of this antibody detected F-actin-capping protein subunit alpha-1 on 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library