Skip to Content
MilliporeSigma

AB5925

Anti-Protein Gene Product 9.5 Antibody

serum, Chemicon®

Synonym(s):

PGP 9.5, UCH-L1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
19
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, pig, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Immunogen

Synthetic peptide corresponding to amino acids 108-127 of soluble cytoplasmic human PGP9.5.

Application

Detect Protein Gene Product 9.5 using this Anti-Protein Gene Product 9.5 Antibody validated for use in WB, IH.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers
Western blot: 1:1,000
Immunohistochemistry: 1:2,000
Optimal working dilutions must be determined by the end user.

Biochem/physiol Actions

Protein Gene Product 9.5 (PGP9.5).

Physical form

Liquid with 0.05% sodium azide.

Preparation Note

Maintain at -20°C in undiluted for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library