Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB5424P

Sigma-Aldrich

Anti-Per1 Antibody

Chemicon®, from rabbit

Synonym(s):

RIGUI, Period Circadian Protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PER1(5187)

Specificity

Recognizes human Per 1 (RIGUI). The immunogen shows no significant sequence homology with Per 2, Per 3, dPer or other known proteins. The immunogen sequence has 100% homology with mouse and rat Per1 and 65% with chicken. Reactivity with other species has not been confirmed. Per 1 is a protein of ~136 kDa.

Immunogen

A 20 amino acid peptide sequence within the C-terminus of human Per1 (Sun et al. 1997; Tei et al. 1997).

Application

Research Category
Neuroscience
Research Sub Category
Circadian Rhythm & Sleep
This Anti-Per1 Antibody is validated for use in ELISA, WB for the detection of Per1.
Western blot: 1-10 μg/mL using ECL.

ELISA: 1:10,000-1:100,000 using 50-100 ng of control peptide (Catalog Number AG330) per well.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS containing 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoqin Liu et al.
PloS one, 7(11), e50602-e50602 (2012-11-29)
Circadian rhythms in metabolism, physiology, and behavior originate from cell-autonomous circadian clocks located in many organs and structures throughout the body and that share a common molecular mechanism based on the clock genes and their protein products. In the mammalian

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service