Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB5390

Sigma-Aldrich

Anti-Sodium Channel Nav1.7 Antibody, Pain

Chemicon®, from rabbit

Synonym(s):

NENA, NE, SCN9A, PN1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

Recognizes Nav1.7 protein (PN1, NENA, NE, Type IX Voltage Gated Sodium Channel, Scn9a). The epitope is specific for Nav1.7 and is not present in any other known protein.

SPECIES REACTIVITIES: The immunogen sequence is highly conserved in rabbit and human (14/15 and 13/15 respectively). Other species have not been tested.

Immunogen

Purified peptide from rat Nav1.7 (amino acids 446-460) (Accession AAB50403).

Application

Detect Sodium Channel Nav1.7 using this Anti-Sodium Channel Nav1.7 Antibody, Pain validated for use in IH & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 1:200 using ECL on rat brain membranes.

Immunohistochemistry on rat dorsal root ganglion (DRG) frozen sections.

Dilutions should be made using a carrier protein such as BSA (1-3%)

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide as a preservative. Reconstitute with 50 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 μL of sterile deionized water. For negative control, preincubate 1 μg of peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui-Yun Bi et al.
PloS one, 12(6), e0178589-e0178589 (2017-06-06)
Temporomandibular disorders (TMDs) have the highest prevalence in women of reproductive age. The role of estrogen in TMDs and especially in TMDs related pain is not fully elucidated. Voltage-gated sodium channel 1.7 (Nav1.7) plays a prominent role in pain perception
Zhongyuan Xia et al.
SpringerPlus, 5(1), 1738-1738 (2016-10-26)
Paclitaxel-induced peripheral neuropathy is not completely known. Since the sodium channel Nav1.7 has been implicated in pain perception, and is upregulated in pain disorders, we investigated the effect of paclitaxel on Nav1.7 expression in rat dorsal root ganglion (DRG) neurons.
R-Y Bi et al.
European journal of pain (London, England), 17(7), 983-994 (2012-12-18)
Inflammation is a major cause of temporomandibular disorder-related pain. The Nav 1.7 sodium channel has a critical function in pain perceptions. However, whether and how Nav 1.7 in the trigeminal ganglion is involved in temporomandibular joint (TMJ) inflammatory pain remains
Rat NaV1.7 loss-of-function genetic model: Deficient nociceptive and neuropathic pain behavior with retained olfactory function and intra-epidermal nerve fibers.
B Grubinska et al.
Molecular pain, 15, 1744806919881846-1744806919881846 (2019-09-26)
Standardized Centella asiatica (ECa 233) extract decreased pain hypersensitivity development in a male mouse model of chronic inflammatory temporomandibular disorder.
Nattapon, et al.
Scientific Reports, 13, 6642-6642 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service