Skip to Content
MilliporeSigma
All Photos(1)

Documents

AB3712

Sigma-Aldrich

Anti-Mannosidase II Antibody, For Golgi Apparatus identification

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

monkey, human

should not react with

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MAN2A1(4124)

Specificity

Mannosidase II. This antibody is a useful marker for golgi apparatus identification.

Immunogen

Epitope: For Golgi Apparatus identification
Purified peptide from human mannosidase II.

Application

Detect the Mannosidase II protein using this Anti-Mannosidase II, For Golgi Apparatus identification validated for use in ICC & IHC.
Immunocytochemistry on HeLa and COS7 cells

Immunohistochemistry on cold methanol fixed tissue.

Optimal working dilutions must be determined by end user.
Research Category
Epitope Tags & General Use
Research Sub Category
Organelle & Cell Markers

Physical form

Rabbit serum. Liquid with 0.02% sodium azide.

Storage and Stability

Maintain unopened vial at -70C for up to 6 months. Avoid repeated freeze/thaw cycles.

The rehydrated antibody solutions can be stored undiluted at 2-8C° for 2 months without any significant loss of activity. Note, the solution is not sterile, thus care should be taken if product is stored at 2-8C°. For storage at -20C°, the addition of an equal volume of glycerol can be used, however, it is recommended that ACS grade or higher glycerol be used, as significant loss of activity can occur if the glycerol used is not of high quality.For freezing, it is recommended that the rehydrated antibody solution be further diluted 1:1 with a 2% BSA (fraction V, highest-grade available) solution made with the rehydration buffer. The resulting 1% BSA/antibody solution can be aliquoted and stored frozen at -70C° for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alex T-W Sin et al.
Molecular and cellular biology, 36(18), 2344-2359 (2016-06-22)
During the cell cycle, genetic materials and organelles are duplicated to ensure that there is sufficient cellular content for daughter cells. While Golgi growth in interphase has been observed in lower eukaryotes, the elaborate ribbon structure of the mammalian Golgi
Yuta Amagai et al.
Nature communications, 14(1), 2683-2683 (2023-05-10)
Many secretory enzymes acquire essential zinc ions (Zn2+) in the Golgi complex. ERp44, a chaperone operating in the early secretory pathway, also binds Zn2+ to regulate its client binding and release for the control of protein traffic and homeostasis. Notably
Azumi Yoshimura et al.
Cells, 11(1) (2022-01-12)
In the early secretory pathway, the delivery of anterograde cargoes from the endoplasmic reticulum (ER) exit sites (ERES) to the Golgi apparatus is a multi-step transport process occurring via the ER-Golgi intermediate compartment (IC, also called ERGIC). While the role
Laasya Dhandapani et al.
Journal of cell science, 135(1) (2021-12-14)
Oocytes spend the majority of their lifetime in a primordial state. The cellular and molecular biology of primordial oocytes is largely unexplored; yet, it is necessary to study them to understand the mechanisms through which oocytes maintain cellular fitness for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service