Skip to Content
MilliporeSigma

AB3442

Anti-CREB Antibody, pSer133

serum, Chemicon®

Synonym(s):

CREB-1, cAMP-response Element-binding Protein-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, monkey, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CREB1(1385)

Immunogen

Epitope: pSer133
Phosphopeptide conjugated to KLH.

Application

Anti-CREB Antibody, pSer133 is a Rabbit Polyclonal Antibody for detection of CREB also known as CREB-1, cAMP-response Element-binding Protein-1 & has been validated in WB.
Immunoblotting: 1:500-1:1,000

Optimal working dilutions must be determined by the end user.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

RNA Binding Protein (RBP)

Biochem/physiol Actions

Recognizes phosphorylated Serine133 in cAMP response element binding protein (CREB). By immunoblotting the antibody shows a 5-10 fold increase in staining of a 43 kDa protein after treatment of cells with agents that increase intracellular cAMP. The antibody also recognizes phosphorylated ATF-1 due to a common epitope. The antibody has been preabsorbed with unphosphorylated peptide and will prove to be useful for the investigation of CREB phosphorylation.

Physical form

Rabbit serum. Liquid. The antibody has been preabsorbed with unphosphorylated peptide. Contains 0.01% sodium azide.

Preparation Note

Maintain at -20°C in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze-thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library