Skip to Content
MilliporeSigma

AB3430

Anti-Prosurfactant Protein B Antibody, reacts with both CT and NT

serum, Chemicon®

Synonym(s):

proSP-B

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA, IHC, WB
Citations:
16
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SFTPB(6439)

Immunogen

Epitope: reacts with both C-terminus and N-terminus
Full length recombinant peptide (381 a.a.) of the human SP-B (pre)proprotein.

Application

Detect Prosurfactant Protein B using this Anti-Prosurfactant Protein B Antibody, reacts with both C-terminus & N-terminus validated for use in ELISA, IH & WB.
Immunohistochemistry: 1:4,000-1:8,000 on human tissue with and without antigen retrieval (see protocol on back) and 1:1,000-1:2,000 on adult and fetal mouse lung (see notes on back).

Immunoblot: 1:1,000-1:5,000; proprotein 42-46kDa.

ELISA: 1:500-1:1,000

Optimal dilutions must be determined by the end user.
Research Category
Metabolism
Research Sub Category
Ion & Transport Channels

Biochem/physiol Actions

Reacts strongly with human and mouse proSP-B (detects both the N-terminal and C-terminal propeptides). Does not react with mature SP-B. On human tissue immunostains alveolar Type II cells and non-ciliated bronchiolar cells. On mouse tissue immunostains alveolar Type II cells and non-ciliated bronchiolar (Clara cells) in distal and terminal airways.

Physical form

Rabbit serum. Liquid. Contains no preservative.

Preparation Note

Maintain at -20°C in undiluted aliquots for up to twelve months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library