Skip to Content
MilliporeSigma

AB3254

Anti-HA Epitope Tag (Influenza Hemaglutinin) Antibody

Chemicon®, from chicken

Synonym(s):

Influenza Hemaglutinin tag

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IP, WB
Citations:
11
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

chicken

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

vertebrates

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Immunogen

Synthetic peptide YPYDVPDYA from influenza hemaglutinin (HA) epitope.

Application

Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags
This Anti-HA Epitope Tag (Influenza Hemaglutinin) Antibody, synthetic peptide YPYDVPDYA is validated for use in IP, WB, IC for the detection of HA Epitope Tag (Influenza Hemaglutinin).
Western blotting (ECL): 1:500

Immunocytochemistry: 1:250

Immunoprecipitation: 1:250

Optimal working dilutions must be determined by end user.

Biochem/physiol Actions

HA (Influenza Hemaglutinin) epitope

Physical form

Affinity purified chicken IgY. Liquid in PBS with 0.02% sodium azide.

Preparation Note

Maintain at 2-8°C in undiluted aliquots, under sterile conditions, for up to 6 months. Freezing the antibody will result in loss of activity of approximately 50%.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 07-221

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library