Skip to Content
MilliporeSigma

AB3252

Anti-c-Myc Tag Antibody

CHEMICON®, chicken polyclonal

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Clone:
polyclonal
Species reactivity:
human
Application:
ICC, IP, WB
Citations:
5
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-c-myc Antibody, Chemicon®, from chicken

biological source

chicken

Quality Segment

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

Immunogen

Synthetic peptide EQKLISEEDL from human c-myc.

Application

Anti-c-myc Antibody is a Chicken Polyclonal Antibody for detection of c-myc also known as Proto-oncogene c-Myc & has been tested in IP, WB, ICC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Western blotting (ECL): 1:500

Immunocytochemistry: 1:250

Immunoprecipitation: 1:250

Optimal working dilutions must be determined by end user.

Biochem/physiol Actions

c-myc

Physical form

Affinity purified chicken IgY. Liquid in PBS with 0.02% sodium azide.

Preparation Note

Maintain at 2-8°C in undiluted aliquots, under sterile conditions, for up to 6 months. Freezing the antibody will result in loss of activity of approximately 50%.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Explore all of our products under Anti-c-myc Antibody

or

Try our Product Selector Tool to narrow your options


wgk

WGK 2

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library