Skip to Content
MilliporeSigma
All Photos(1)

Documents

AB3203

Sigma-Aldrich

Anti-Tubulin Antibody, δ 2

Chemicon®, from rabbit

Synonym(s):

Anti-α-tubulin, Tubulin antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

sea urchin, echinoderm, plant

should not react with

Saccharomyces cerevisiae

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TUBD1(51174)

General description

Tubulin, the dimeric subunit of microtubules, is submitted to post translational modifications including cyclic tyrosine removal and addition at the COOH terminus of the alpha subunit. The cycle involves two enzymes, TTL (Tubuline Tyrosine Ligase) and Tubulin carboxypeptidase, and generates two major forms: tyrosinated tubulin and detyrosinated tubulin (Glu-tubulin). Removal of the COOH-terminal Glu residue from the alpha chain of Glu-tubulin generates the delta2-tubulin form.

Specificity

The antibody specifically recognizes the delta 2 form of the tubulin alpha chain (Paturle-Lafanechere et al. 1991).

SPECIES REACTIVITY:

Mammals, sea urchin, plants, not yeast (Lafanechere et al. 1994).

Immunogen

Peptide corresponding to the C-terminal of delta 2 tubulin.

Application

Research Category
Cell Structure
Research Sub Category
Cytoskeleton
This Anti-Tubulin Antibody, δ 2 is validated for use in IH(P), WB, IC, IH for the detection of Tubulin.
Western blot: 1:1,000

Immunohistochemistry and EM on either frozen or paraffin embedded, fixed tissue (Mialhe et al. 2001): 1:500

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS.

Storage and Stability

Maintain at 2-8°C in undiluted for up to 6 months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert O'Hagan et al.
Current biology : CB, 21(20), 1685-1694 (2011-10-11)
Posttranslational modifications (PTMs) such as acetylation, detyrosination, and polyglutamylation have long been considered markers of stable microtubules and have recently been proposed to guide molecular motors to specific subcellular destinations. Microtubules can be deglutamylated by the cytosolic carboxypeptidase CCP1. Loss
Kinesin-13 and tubulin posttranslational modifications regulate microtubule growth in axon regeneration.
Ghosh-Roy, A; Goncharov, A; Jin, Y; Chisholm, AD
Developmental Cell null
Post-translational modifications to Toxoplasma gondii alpha- and beta-tubulins include novel C-terminal methylation.
Xiao, H; El Bissati, K; Verdier-Pinard, P; Burd, B; Zhang, H; Kim, K; Fiser, A; Angeletti et al.
Journal of Proteome Research null
Hung-Chi Liu et al.
Plant physiology, 191(1), 177-198 (2022-10-23)
Phosphoglucose isomerase (PGI) catalyzes the interconversion of fructose-6-phosphate and glucose-6-phosphate, which impacts cell carbon metabolic flow. Arabidopsis (Arabidopsis thaliana) contains two nuclear PGI genes respectively encoding plastidial PGI1 and cytosolic PGI (cPGI). The loss of PGI1 impairs the conversion of
Dynamics of DNA methylation and Histone H4 acetylation during floral bud differentiation in azalea.
Meijon, M; Feito, I; Valledor, L; Rodriguez, R; Ca?al, MJ
BMC plant biology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service