Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB3202

Sigma-Aldrich

Anti-LIM-3 Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-HMFN1661

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

fish, frog, mouse, human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ABLIM3(22885)

Specificity

Recognizes LIM-3. This antibody does not appear to cross react with gsh-4 on tissue sections but may cross react in Western blot and immunoprecipitation.

Immunogen

C-terminal portion of mouse LIM-3 protein.

Application

Anti-LIM-3 Antibody is an antibody against LIM-3 for use in IP, WB, IC, IH, IH(P).
Immunohistochemistry: 1:250-1:1,000 on paraffin embedded sections. 1:3,000-1:6,000 on frozen sections. 1:4,000-1:8,000 on whole mounts. Recommended fixative MEMFA.

Immunocytochemistry: 1:200-1:500

Western blot: 1:2,500-1:5,000

Immunoprecipitation: 1:200

This antibody will work well in immunofluorescence. Use a secondary antibody conjugated to biotin and then streptavidin-conjugated to fluorochrome.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Neuronal & Glial Markers

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02M Phosphate buffer, 0.25M NaCl with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mai Nakamura et al.
Cytotechnology, 68(3), 409-417 (2014-10-31)
Mouse embryonic stem (ES) cells and induced pluripotent stem (iPS) cells have the ability to differentiate in vitro into various cell lineages including neurons. The differentiation of these cells into neurons has potential applications in regenerative medicine. Previously, we reported
Clifford R Hume et al.
Gene expression patterns : GEP, 7(7), 798-807 (2007-07-03)
A cascade of transcription factors is believed to regulate the coordinate differentiation of primordial inner ear cells into the subtypes of hair cells and supporting cells. While candidate genes involved in this process have been identified, the temporal and spatial
Stephanie W Fowler et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(23), 7871-7885 (2014-06-06)
An unresolved debate in Alzheimer's disease (AD) is whether amyloid plaques are pathogenic, causing overt physical disruption of neural circuits, or protective, sequestering soluble forms of amyloid-β (Aβ) that initiate synaptic damage and cognitive decline. Few animal models of AD
Anne-Laure Schang et al.
Molecular endocrinology (Baltimore, Md.), 27(1), 74-91 (2012-12-06)
GATA2 transcription factor and LIM homeodomain proteins Islet1 (ISL1) and LIM homeobox 3 (LHX3) are suspected to be involved in gonadotrope cell fate and maintenance. The GnRH receptor gene (Gnrhr), crucial for gonadotrope function, is expressed in the pituitary gland
Zhiyong Liu et al.
PloS one, 9(2), e89377-e89377 (2014-03-04)
Regeneration of auditory hair cells (HCs) is a promising approach to restore hearing. Recent studies have demonstrated that induced pluripotent stem cells/embryonic stem cells or supporting cells (SCs) adjacent to HCs can be converted to adopt the HC fate. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service