Skip to Content
MilliporeSigma

AB3116

Anti-Bcl-xL Antibody, Phospho-specific

serum, Chemicon®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... BCL2L1(598)

General description

Bcl-XL is one of the anti-apoptotic members of the Bcl-2 family of apoptosis regulators and is overexpressed in many human cancers. Studies by Dr. Haldar and his associates suggest that microtubule damaging drugs like Taxol or 2-methoxyestradiol induce phosphorylation of Bcl-XL at serine 62 by kinases such as Jun and this allows those cells to die by apoptosis.

Immunogen

Epitope: Phospho-specific
Synthetic phosporylated peptide corresponding to a sequence encompassing serine 62 of human Bcl-xL conjugated to KLH.

Application

Detect Bcl-xL using this Anti-Bcl-xL Antibody, Phospho-specific validated for use in WB.
Immunoblotting: Recommended dilution is 1:100

Optimal dilutions must be determined by end-user.
Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition

Tumor Markers

Biochem/physiol Actions

AB3116 is specific for the Bcl-xL phosphorylated on serine 62. It does not recognize the unphosphorylated form of the protein.

Physical form

Liquid rabbit serum containing no preservatives.

Preparation Note

Maintain at -80°C for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library