Skip to Content

AB3098

Anti-Orexin-A Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-NRCLP1, Anti-OX, Anti-PPOX

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA, WB
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

feline, rat, human, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PPOX(5498)

Immunogen

An 18 amino acid peptide mapping near the amino terminus of mouse Orexin-A. The immunogen sequence is identical in human, rat, porcine, ovine, bovine and canine and has 81% homology with chicken.

Application

Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Circadian Rhythm & Sleep
This Anti-Orexin-A Antibody is validated for use in ELISA, WB for the detection of Orexin-A.
Western blot: 1-10 μg/mL using Chemiluminescence technique ELISA: 1:10,000 - 100,000 using 5-100 ng of control peptide per well. NOTE: Orexin A is a 3.5kDa protein. Adjust your SDS-PAGE gel and lane size markers accordingly.

Immunohistochemistry: Frozen 4% PFA tissue with ABC detection {Wagner et al 2000}.

Optimal working dilutions must be determined by end user.

Biochem/physiol Actions

Mouse Orexin-A. Several peptides associated with feeding behavior have been reported recently. Orexins (Orexin-A and Orexin-B) are a family of hypothalamic neuropeptides selectively expressed in the hypothalamus (Sakurai et al. 1998). Orexin-A and Orexin-B are derived from the same precursor (Prepro-orexin) by proteolytic cleavage. Prepro-orexin is an 130 aa peptide with a putative 33 aa secretory sequence, a hydrophobic core followed by residues with small polar side chains. The expression was detected in brain and to a small extent in testis (Sakurai et al. 1998). These neuropeptides bind and activate two closely related Orexin receptors--G protein coupled receptors (GPCRs) OX1R and OX2R.

Physical form

Liquid in PBS, pH 7.4 with 0.1% BSA and 0.1% sodium azide.

Preparation Note

Maintain frozen at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Explore all of our products under Anti-Orexin-A Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library