Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB3000

Sigma-Aldrich

Anti-IP3 Receptor Type II Antibody

Chemicon®, from rabbit

Synonym(s):

Inositol 1,4,5-Trisphosphate Receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, hamster, human, monkey, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITPR2(3709)

Specificity

Rabbit anti-Type II Inositol 1,4,5-Trisphosphate Receptor (IP3R) is directed against a synthetic peptide corresponding to the carboxy terminus of the rat Type II receptor. The antibody is reactive with Type II IP3R from rat, mouse, human, hamster, and monkey.

Immunogen

Synthetic peptide corresponding to the carboxy terminus of the rat Type II receptor

Application

Anti-IP3 Receptor Type II Antibody detects level of IP3 Receptor Type II & has been published & validated for use in IC, IF, IH, IP & WB.
Research Category
Signaling
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling
Western blot: 1:50 - 1:100 (detects a 260 kDa band)

Immunofluorescence: 1:10 - 1:20

Immunoprecipitation

Immunocytochemistry: Known to give positive reactivity on AR4-2J, HL-60, COS-I, HEK-293, and RBL-2H3 cell lines.

Immunohistochemistry: Paraformaldehyde or methanol fixes are effective.

Optimal working dilutions must be determined by end user.

Physical form

Affinity purified versus immobilized immunogen peptide. Liquid in PBS buffer, containing 0.05% BSA, 10% glycerol, 0.005% sodium azide. Caution: Sodium azide is hazardous. MSDS available upon request.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark Chalmers et al.
The Biochemical journal, 394(Pt 1), 57-66 (2005-11-09)
The size and number of IP3R (inositol 1,4,5-trisphosphate receptor) clusters located on the surface of the ER (endoplasmic reticulum) is hypothesized to regulate the propagation of Ca2+ waves in cells, but the mechanisms by which the receptors cluster are not
Robert P Brumer et al.
The Journal of physiology, 601(8), 1383-1405 (2023-03-04)
Excess consumption of carbohydrates, fat and calories leads to non-alcoholic fatty liver disease (NAFLD) and hepatic insulin resistance; these are major factors in the pathogenesis of type II diabetes. Hormones and catecholamines acting through G-protein coupled receptors (GPCRs) linked to
Shafi Kuchay et al.
Nature, 546(7659), 554-558 (2017-06-15)
In response to environmental cues that promote IP3 (inositol 1,4,5-trisphosphate) generation, IP3 receptors (IP3Rs) located on the endoplasmic reticulum allow the 'quasisynaptical' feeding of calcium to the mitochondria to promote oxidative phosphorylation. However, persistent Ca2+ release results in mitochondrial Ca2+
Ji-Wei Li et al.
Oncotarget, 7(40), 65849-65861 (2016-09-03)
Oxsterol binding protein-related protein 4 (ORP4) is essential for cell proliferation, but the underlying mechanism is unclear. ORP4 is expressed as three variants, ORP4L, ORP4M and ORP4S. Here, we reported that silencing of ORP4L with specific small interfering RNA (siRNA)
Takahiro Fujimoto et al.
International journal of molecular medicine, 30(6), 1287-1293 (2012-09-21)
KRAS-induced actin-interacting protein (KRAP), originally identified as one of the deregulated genes expressed in colorectal cancer, participates under physiological conditions in the regulation of systemic energy homeostasis and of the exocrine system. We have recently found that KRAP is a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service