Skip to Content

AB2218

Anti-Aquaporin-4 Antibody

CHEMICON®, rabbit polyclonal

Synonym(s):

AQP4, WCH4, MIWC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Clone:
polyclonal
Species reactivity:
human, mouse, rat
Application:
WB
Citations:
24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-Aquaporin 4 Antibody, Chemicon®, from rabbit

biological source

rabbit

Quality Segment

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AQP4(361)

General description

Aquaporin-4 (UniProt: P47863; also known as AQP-4, Mercurial-insensitive water channel (MIWC), WCH4) is encoded by the Aqp4 gene (Gene ID: 25293) in rat. Aquaporin-4 is a transmembrane water channel protein predominantly expressed in astrocytes within the central nervous system (CNS), where it plays a critical role in maintaining water homeostasis and supporting key physiological processes. Strategically localized at the perivascular end-feet of astrocytes, Aquaporin-4 facilitates bidirectional water transport across the blood-brain barrier and is integral to the glymphatic system, a clearance pathway responsible for removing interstitial solutes such as amyloid-beta. Dysregulation of Aquaporin-4 has been implicated in numerous neuropathological conditions, including neuromyelitis optica spectrum disorder (NMOSD), Alzheimer′s disease, traumatic brain injury, and brain edema. In NMOSD, autoantibodies targeting Aquaporin-4 lead to astrocyte destruction, inflammation, and central nervous system damage, emphasizing the importance of early and accurate detection methods such as biosensors. Aquaporin-4 isoforms, particularly M23, form supramolecular structures called orthogonal arrays of particles (OAPs), which are crucial for its localization and function. Extended isoforms such as AQP4ex further regulate water channel activity and contribute to amyloid-beta clearance, highlighting their therapeutic potential in neurodegenerative diseases. Redistribution of Aquaporin-4 within astrocytes, often due to disruptions in its anchoring dystrophin-associated protein complex, has been associated with impaired glymphatic function, exacerbating conditions like idiopathic normal pressure hydrocephalus and glioblastoma. (Ref.: Silverglate, B., et al., (2023), Expert Opin.Therap. Targets, 27(7), 523-530; Peng, S., et al., (2023), Neurobiol. Dis. 179, Article 106035; Vandebroek, A., and Yasui, M. (2020), Int. J. Mol. Sci. 21(5); 1603).
Target molecular weight ~32 kDa observed. 34.48 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal cytoplasmic domain of human.

Application

Anti-Aquaporin-4, Cat. No. AB2218 is a rabbit polyclonal antibody that detects Aquaporin-4 and is tested for use in Western Blotting and Immunohistochemistry (Paraffin).
Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Aquaporin-4 in rat brain tissue lysate.




Tested applications

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Aquaporin-4 in human and mouse brain tissue lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Aquaporin-4 in human, rat, and mouse brain tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Research Category
Neuroscience

Biochem/physiol Actions

This rabbit polyclonal antibody detects Aquaporin-4. It targets an epitope within 16 amino acids from the C-terminal cytoplasmic domain.

Physical form

Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Recommended storage: +2°C to +8°C.

Analysis Note

Control
Tested on adult rat brain lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: AB3068

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Explore all of our products under Anti-Aquaporin 4 Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library