Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.43
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
5
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... CCR5(1234)
General description
HIV and related viruses require coreceptors, in addition to CD4, to infect target cells. Some G-protein-coupled receptors including CCR5, CXCR4, CCR3, CCR2b and CCR8 in the chemokine receptor family, and four new human molecules GPR15, STRL33, GPR1 and V28 were recently identified as HIV coreceptors (1,2). Among them, CCR5 is a principle coreceptor for macrophage- and dual-tropic HIV-1 strains fusion of entry of human white blood cells (3,4). The amino-terminal domain and the extracellular loops of CCR5 serve as HIV binding sites. CCR5 messenger RNA is expressed in lymphoid organs and monocytes.
Immunogen
Epitope: N-terminus
Synthetic peptide from the N-terminal domain of human CCR5.
Application
Anti-C-C Chemokine Receptor 5 Antibody, N-terminus is an antibody against C-C Chemokine Receptor 5 for use in WB.
Research Category
Inflammation & Immunology
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors
Cytokines & Cytokine Receptors
Western blot: 1:1000 - 1:2000 (THP-1 whole cell lysate can be used as a positive control)
Optimal working dilutions must be determined by end user.
Optimal working dilutions must be determined by end user.
Biochem/physiol Actions
Specific for human C-C Chemokine Receptor 5 (CCR5) at the N-terminus.
Physical form
Format: Purified
Purified immunoglobulin. 100 μg purified IgG in 100 μL of PBS containing 0.2% sodium azide.
Preparation Note
Store at 2-8°C for up to 12 months.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.