Skip to Content

AB1870

Anti-Osteopontin Antibody

serum, Chemicon®

Synonym(s):

Uropontin, Bone sialoprotein 1, SPP1/CALPHA1 fusion, Urinary stone protein, bone sialoprotein I, osteopontin/immunoglobulin alpha 1 heavy chain constant region fusion protein, secreted phosphoprotein 1, secreted phosphoprotein 1 (osteopontin, bone sialop

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA, IHC (p), WB
Citations:
30
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SPP1(6696)

General description

Osteopontin is a single-chain polypeptide with a molecular weight of approximately 35 kDa. It is a glycoprotein first identified in osteoblasts. It is biosynthesized by a variety of tissue types including preosteoblasts, osteoblasts, osteocytes, extraosseous cells in the inner ear, brain, kidney, deciduum, placenta, odontoblasts, some bone marrow cells, hypertrophic chondrocytes, macrophages, smooth muscle, and endothelial cells. Osteopontin is overexpressed in a variety of cancers, including lung cancer, breast cancer, colorectal cancer, stomach cancer, ovarian cancer, melanoma and mesothelioma. It may contribute to kidney stone formation and both glomerulonephritis and tubulointerstitial nephritis and is also found in atheromatous plaques within arteries. Research has implicated osteopontin in excessive scar-forming and a gel has been developed to inhibit its effect.
~35 kDa

Immunogen

Synthetic peptide corresponding to amino acids 75-90 of human osteopontin (SwissProt Accession # P10451; PSKSNESHDHMDDM DD).

Application

Immunohistochemistry:
A previous lot of this antibodu was used at a 1:1500 dilution on paraffin-embedded tissue sections.

ELISA:
A previous lot of this antibody was used at a 1:1000 dilution.

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Osteobiology
This Anti-Osteopontin Antibody is validated for use in ELISA, IH, IH(P), WB for the detection of Osteopontin.

Biochem/physiol Actions

Expected to cross-react with rat.
Human osteopontin. Does not cross react with osteonectin or human bone sialoprotein II.

Physical form

Rabbit polyclonal antiserum in buffer containing no preservatives.
Unpurified

Preparation Note

Stable for 1 year at -20°C in undiluted aliquots from date of receipt.

Handling Recommendations:
Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
POSITIVE CONTROL: Human urine (2-5 µg/mL normal concentration).
Routinely evaluated by Western Blot on RAW 264.7 lysates.

Western Blot Analysis: 1:500 dilution of this lot detected osteopontin on 10 μg of RAW 264.7 lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Explore all of our products under Anti-Osteopontin Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library