Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB1280

Sigma-Aldrich

Anti-Cytochrome P450 Enzyme CYP4A1/2/3 Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

Binds specifically to cytochrome P450 CYP4A1, CYP4A2 and CYP4A3 in rat hepatic microsomal fraction.

Immunogen

Synthetic peptide

Application

Anti-Cytochrome P450 Enzyme CYP4A1/2/3 Antibody is an antibody against Cytochrome P450 Enzyme CYP4A1/2/3 for use in IH & WB.
Research Category
Metabolism
Research Sub Category
Enzymes & Biochemistry
Western blot: hepatic microsomal membrane preps recommended; ~55/58 kDa on reducing gels

Immunohistochemistry: 4% PFA fixed, frozen tissue recommended.

Optimal working dilutions must be determined by end user.

Physical form

Rabbit antiserum. Liquid, no preservatives.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service