Skip to Content
MilliporeSigma

AB10140

Anti-Drebrin A/E Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Developmentally-regulated brain protein, drebrin 1, drebrin E, drebrin E2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
3
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

technique(s)

immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DBN1(1627)

General description

The developmentally-regulated neuron-specific protein, Drebrin A, is expressed first at the time of outgrowth and maturation of dendrites, and is localized within dendrites of the adult brain. Drebrin E+A- signals have been observed in the cell bodies of migrating neuroblasts in the core region; however, only fibrous and punctate drebrin E+A- signals were observed in postmigratory neuroblasts at the outer layers. Studies have demonstrated that the disappearance of drebrin E+A- signals from the cell body coincides with the cessation of neuronal migration. The disappearance of drebrin E from the cell body may be a molecular switch for the cessation of migration in newly generated neuroblast
~77 kDa

Immunogen

Epitope: Internal Domain
KLH conjugated synthetic linear peptide.

Application

Detect Drebrin A/E using this Anti-Drebrin A/E Antibody validated for use in WB & IC.
Immunocytochemistry: Proven to react with Drebrin A/E in 16 DIV rat cortical neurons. Photo courtesy of Dr. Gang Lui, Drexel University.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology

Biochem/physiol Actions

Cat. # AB10140 detects the internal domain of Drebrin A/E.
May react with human based on sequence homology. Reactivity with other species has not been determined.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal antibody in PBS with 0.05% NaN3.

Preparation Note

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
Embryonic rat brain (day 16) tissue lysate.
Western Blot:
A 1:5,000 dilution of this antibody was used to detect embryonic rat brain (day 16) tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library