Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.56114

Sigma-Aldrich

Fmoc-Met-Wang resin LL (100-200 mesh)

Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-30°C

General description

Pre-loaded resin for synthesis of peptide acids containing a C-terminal methionine amino-acid residue by Fmoc SPPS. Prepared from a low load Wang resin, to help facilitate the synthesis of longer peptides.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Linkage

Replaces: 04-12-2084

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.20 - 0.40 mmol/g
Enantiomeric purity: lot specific result
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly (styrene-1 % DVB), 100 - 200 mesh. The resin is endcapped after coupling the amino acid derivative.
The polymer matrix is copoly (styrene-1 % DVB), 100 - 200 mesh. The resin is endcapped after coupling the amino acid derivative.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service