Skip to Content
MilliporeSigma

8.56054

H-Lys(Boc)-2-ClTrt resin

Novabiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.22
UNSPSC Code:
12352209
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

General description

Pre-loaded resin for synthesis of protected peptide acids containing a C-terminal lysine amino-acid residue by Fmoc SPPS. Cleavage of the protected peptide is effected by treatment with 1% TFA in DCM or 20% TFE in DCM

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.30 - 0.90 mmol/g
Swelling Volume (in DMF): lot specific result
Identity (of the substitution): passes test
The polymatrix is copoly (styrene -1% DVB ), 200 - 400 mesh.

Other Notes

Replaces: 04-12-2802

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library