Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.56044

Sigma-Aldrich

Fmoc-Cys(Trt)-NovaSyn® TGT

for peptide synthesis, Novabiochem®

Synonym(s):

Trityl-Protected Cysteine Resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352209
NACRES:
NA.22

product name

Fmoc-Cys(Trt)-NovaSyn® TGT, Novabiochem®

Quality Level

product line

NovaSyn® TG
Novabiochem®

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

15-25°C

General description

Pre-loaded resin for synthesis of peptide acids and protected peptide fragments containing a C-terminal cysteine amino-acid residue by Fmoc SPPS. The base NovaSyn® TG is a composite of low cross-linked polystyrene and 3000-4000 M.W. polyethylene glycol. Peptide synthesis is carried out at the ends of the PEG chains that have been functionalized with the hyper-acid labile 4-carboxytrityl alcohol linker. This use of this linker helps prevent racemization and β-piperidinylalanine formation during chain extension.Treatment of the peptidyl resin with 20% TFE in DCM or 1% TFA in DCM cleaves the product from the resin without affecting the standard TFA-labile side-chain protecting groups. Standard TFA cleavage releases the fully deprotected peptide.

Associated Protocols and Technical Articles:
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references:
[1] E. Atherton, et al. in "Peptides 1990, Proc. 21st European Peptide Symposium, E. Giralt & D. Andreu(Eds), 1991, Escom, Leiden, pp. 243
[2] J. Lukszo, et al. (1996) Lett. Pept. Sci., 3, 157.
[3] Y. Fujiwara, et al. (1994) Chem. Pharm. Bull., 42, 724.
[4]
K. Barlos & D. Gatos in “Fmoc solid phase peptide synthesis: a practical approach”, W. C. Chan & P. D.White (Eds.), Oxford University Press, Oxford, 2000, pp. 218.

Application

Applications of Fmoc-Cys(Trt)-NovaSyn® TGT include:
  • the preparation of a phosphoprotein using Native Chemical Ligation (NCL).
  • the preparation of a peptide intermediate for use in glycoprotein semi-synthesis.

Linkage

Replaces: 04-12-2705

Legal Information

NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Barlos & D. Gatos in ?Fmoc solid phase peptide synthesis: a practical approach?, W. C. Chan & P. D. White (Eds.), Oxford University Press, Oxford
K. Barlos & D. Gatos
Solid Phase Peptide Synthesis: A Practical Approach, 218-218 (2000)
3-(1-Piperidinyl) alanine formation during the preparation of C-terminal cysteine peptides with the Fmoc/t-Bu strategy
J. Lukszo, et al.
Peptide science (Hoboken, N.J.), 3, 157-157 (1996)
Peptides 1990, Proc. 21st European Peptide Symposium, E. Giralt & D. Andreu
E. Atherton, et al. in
Proceedings, 243-243 (1991)
Access to phosphoproteins and glycoproteins through semi-synthesis, Native Chemical Ligation and N? S acyl transfer
Masania J, et al.
Organic & Biomolecular Chemistry, 5113-5119 (2010)
Racemization-free synthesis of C-terminal cysteine-peptide using 2-chlorotrityl resin
Y. Fujiwara, et al.
Journal of Chemical and Pharmaceutical Sciences , 42, 724-724 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service