Skip to Content
MilliporeSigma

8.55102

Ethyl Indole AM resin

Novabiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.22
UNSPSC Code:
12352005
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

aldehyde

storage temp.

2-8°C

General description

[3-((Ethyl-Fmoc-amino)-methyl)indol-1-yl] acetyl AM resin is an extremely useful support for the production of peptide N-ethylamides, N-ethyl substituted carboxamides and sulfonamides [1]. After first removing the Fmoc group, loading of the resin with Fmoc-amino acids can be effected using HATU/DIPEA. Cleavage with TFA directly provides the product N-ethylamide.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] K. G. Estep, et al. (1998) J. Org. Chem., 63, 5300.

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.80 - 1.10 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly (styrene-1% DVB), 100 - 200 mesh

Other Notes

Replaces: 01-64-0398

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library