Skip to Content
MilliporeSigma

8.55074

Weinreb AM resin

Novabiochem®

Synonym(s):

N-Fmoc- N-methoxy-β-alanine AM resin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.22
UNSPSC Code:
12352203
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-30°C

General description

This support is useful for the production of peptide aldehydes and other carboxaldehydes. Following removal of the Fmoc group with 20% piperidine in DMF, acylation of the resin-bound methoxylamine is best effected using DIPCDI/HOAt or HATU/DIPEA activation. This results in formation of a supported Weinreb-type amide which can be either reduced to an aldehyde with LiAlH4 [1,2] or cleaved with Grignard reagents to give ketones [2,3].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.40 - 1.00 mmol/g
Swelling Volume (in CH₂Cl₂): lot specific result
The polymer matrix is copoly (styrene - 1 % DVB), 100 - 200 mesh

Other Notes

Replaces: 01-64-0153

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library