Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.53048

Sigma-Aldrich

Boc-Asp(OtBu)-OH . DCHA

Novabiochem®

Synonym(s):

Boc-Asp(OtBu)-OH . DCHA, N-α-t.-Boc-L-aspartic acid β-t.-butyl ester dicyclohexylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H23NO6 · xC12H23N
CAS Number:
Molecular Weight:
289.32
MDL number:
UNSPSC Code:
12352209
EC Index Number:
217-626-7
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥98% (TLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

carboxylic acid

storage temp.

2-30°C

SMILES string

[N+H2](C2CCCCC2)C1CCCCC1.N([C@@H](CC(=O)OC(C)(C)C)C(=O)[O-])C(=O)OC(C)(C)C

InChI

1S/C13H23NO6.C12H23N/c1-12(2,3)19-9(15)7-8(10(16)17)14-11(18)20-13(4,5)6;1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h8H,7H2,1-6H3,(H,14,18)(H,16,17);11-13H,1-10H2/t8-;/m0./s1

InChI key

OMYRDWOMNDCKEJ-QRPNPIFTSA-N

General description

Building block of introduction of N-terminal aspartic acid amino-acid residues by Fmoc SPPS

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS

Linkage

Replaces: 04-12-0040

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in methanol): +15.5 - +19.5 °
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service