Skip to Content
MilliporeSigma

8.53028

Boc-Glu-OtBu

Novabiochem®

Synonym(s):

Boc-Glu-OtBu, N-α-t.-Boc-L-glutamic acid α-t.-butyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H25NO6
CAS Number:
Molecular Weight:
303.35
UNSPSC Code:
12352209
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥98% (TLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

carboxylic acid

storage temp.

2-30°C

SMILES string

N(C(CCC(=O)O)C(=O)OC(C)(C)C)C(=O)OC(C)(C)C

InChI

1S/C14H25NO6/c1-13(2,3)20-11(18)9(7-8-10(16)17)15-12(19)21-14(4,5)6/h9H,7-8H2,1-6H3,(H,15,19)(H,16,17)

InChI key

YMOYURYWGUWMFM-UHFFFAOYSA-N

General description

Derivative of synthesis of side-chain modified glutamic acid derivatives.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in methanol): -32.5 - -27.5 °
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (1 mmole in 2 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-0111

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Boc-Glu-OtBu


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library